Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-N-methyl-D-phenylalanine, 95%

Catalog No. AAH63055MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63055MD 250 mg
AAH6305503 1 g
AAH6305506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63055MD Supplier Thermo Scientific Chemicals Supplier No. H63055MD

CAS: 138775-05-0 | C25H23NO4 | 401.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138775-05-0
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00153392
InChI Key GBROUWPNYVBLFO-HSZRJFAPSA-N
Synonym fmoc-n-methyl-d-phenylalanine, fmoc-d-mephe-oh, fmoc-n-me-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-nalpha-methyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, ambotzfaa1333, fmoc-nme d phe-oh, fmoc-n-, a-methyl-d-phenylalanine
PubChem CID 978357
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Beilstein 8441627
Quantity 250 mg
Formula Weight 401.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-D-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.