Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-ethyl-L-cysteine, 95%, Thermo Scientific™

Catalog No. AAH6601506
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6601506 5 g
AAH66015MD 250 mg
AAH6601503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6601506 Supplier Thermo Scientific Chemicals Supplier No. H6601506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200354-34-3
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.451
MDL Number MFCD00151921
InChI Key OUXUHWHQQLGDQE-SFHVURJKSA-N
Synonym fmoc-cys et-oh, fmoc-s-ethyl-l-cysteine, l-cysteine,s-ethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-ethylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-ethyl-n-9h-fluoren-9-yl methoxy carbonyl-l-cysteine
PubChem CID 7015833
IUPAC Name (2R)-3-ethylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CCSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 5 g
Formula Weight 371.45
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-S-ethyl-L-cysteine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.