Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formylglycine Ethyl Ester 98.0+%, TCI America™
SDP

Catalog No. F042125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F042125G 25 g
F04215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F042125G Supplier TCI America Supplier No. F042125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3154-51-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00014125
InChI Key GMBCCEOJUWMBPF-UHFFFAOYSA-N
Synonym n-formylglycine ethyl ester, ethyl n-formylglycinate, glycine, n-formyl-, ethyl ester, ethyl formylaminoacetate, ethylformylglycine, n-formylglycineethylester, ethyl formamidoacetate, 2-formylaminoacetic acid ethyl ester, for-gly-oet, ethyl formylglycinate
PubChem CID 76615
IUPAC Name ethyl 2-formamidoacetate
SMILES CCOC(=O)CNC=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Formylglycine Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.