Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Formylmorpholine, 99%

Catalog No. AC133801000
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133801000 100 mL Glass bottle
AC133800050 5 mL Glass bottle
AC133805000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC133801000 Supplier Thermo Scientific Chemicals Supplier No. 133801000
Only null left
Add to Cart

CAS: 4394-85-8 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 23°C
Color Yellow
Density 1.1400g/mL
Boiling Point 240°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4855 to 1.4875
Quantity 100 mL
Specific Gravity 1.14
Solubility Information Solubility in water: soluble in water
Viscosity 6.79 mPa.s (30°C)
Formula Weight 115.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Formylmorpholine, 99+%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 224-518-3
RTECSNumber QD9694000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.