Alfa Aesar™ n-Heptane, 95%

Manufacturer: Alfa Aesar™ H33987AU

Quantity 1000mL

 View more versions of this product

Catalog No. AAH33987AU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142-82-5
    Molecular Formula C7H16
    Linear Formula CH3(CH2)5CH3
    Formula Weight 100.21
    MDL Number MFCD00009544
    UN Number UN1206
    Beilstein 1730763
    Merck Index 14,4659
    Chemical Name or Material n-Heptane

    Specifications

    Formula Weight 100.21
    UN Number UN1206
    Flash Point −4°C (25°F)
    Melting Point −91°C
    Quantity 1000mL
    TSCA TSCA
    Molecular Formula C7H16
    Linear Formula CH3(CH2)5CH3
    MDL Number MFCD00009544
    CAS 142-82-5
    Beilstein 1730763
    Merck Index 14,4659
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
    RTECS Number MI7700000
    Odor Petroleum-like
    Boiling Point 97° to 99°C
    Chemical Name or Material n-Heptane
    Assay 95%

    Description

    Applications
    n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

    Solubility
    Miscible with alcohol, chloroform, and ether. Insoluble in water.

    Notes
    Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.

    GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit