Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane AR, Macron™

Catalog No. MK5177500 Shop All Avantor Products
Change view
Click to view available options
Glass; 500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-5177-500 Glass; 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-5177-500 Supplier Avantor Supplier No. 517704

missing translation for 'validMainframeMsg'

Specifications

CAS 142-82-5
Quantity 500 mL
Packaging Glass
Molecular Formula C7H16
MDL Number MFCD00009544
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
PubChem CID 8900
ChEBI CHEBI:43098
Formula Weight 100.2
Chemical Name or Material n-Heptane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.