Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog No. 01100239 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-239 1 L
01-100-240 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-100-239 Supplier MilliporeSigma Supplier No. 1.03654.1000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Density 0.68 g/cm3 at 15 °C
Quantity 1 L
Explosion Limit 1 to 7% (V)
Flash Point -4 °C
Ignition Point 215 °C
Packaging Glass Bottle
Residue after Evaporation ≤ 2 mg/L
Vapor Pressure 111 hPa at 37.7 °C
Water ≤ 0.005%
Boiling Point 97°C to 98°C (1013 hPa)
Solubility Information 0.05 g/L
Percent Purity ≥99.3%
Grade Hypergrade
Show More Show Less
Recommended Storage Store at 2 to 30 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.