Learn More
Strem, An Ascensus Company CAS# 250285-32-6. 2G. 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, min. 97%. MFCD02684545
SDP
Supplier:  Strem, An Ascensus Company 0705902G
Encompass_Preferred
CAS# 250285-32-6. 2G. 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, min. 97%. MFCD02684545. Molecular Formula: C27H37N2+Clae. Molecular Weight: 425.06. Color/Form: white to off-white pwdr. Strem# 07-0590. www.strem.com/catalog/v3/07-0590/
Catalog No. 50-196-3585
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.