Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
GREENER_CHOICE

Catalog No. AC445720025
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445720025 2.5 L
AC445720010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC445720025 Supplier Thermo Scientific Chemicals Supplier No. 445720025
  • $303.00
    $241.65 / Each of 1

    Save $61.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-54-3, C6H14, 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym gettysolve-b, n-hexane, hexyl hydride, esani, heksan, skellysolve b, hexan, hexanes, dipropyl, hexanen
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
Density 0.6590g/mL
Boiling Point 69.0°C
Flash Point -22°C
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
Refractive Index 1.3748 to 1.3810
Quantity 2.5 L
Specific Gravity 0.659
Merck Index 15,4732
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Percent Purity 97+%
Grade HPLC
Green Features ACT Ecolabel (Silver)
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long las
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or d
EINECSNumber 203-777-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.