Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane-d{14}, 99%(Isotopic)

Catalog No. AA4231203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4231203 1 g
AA4231206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4231203 Supplier Thermo Scientific Chemicals Supplier No. 04231203
Add to Cart

CAS: 21666-38-6 | C6H14 | 100.26 g/mol

n-Hexane-d{14} is a deuterium labeled n-hexane which is used as a organic intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21666-38-6
Molecular Formula C6H14
Molecular Weight (g/mol) 100.26
MDL Number MFCD00037562
InChI Key VLKZOEOYAKHREP-ZLKPZJALSA-N
Synonym hexane-d14, tetradecadeuterohexane, n-hexane-d 14, 2h14 hexane, 2 h?? hexane, hexane-d14, 99 atom % d
PubChem CID 140854
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecadeuteriohexane
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Quantity 1 g
Density 0.767 g/mL
Flash Point −23°C (−9°F)
Odor Characteristic
Melting Point -95°C
Boiling Point 69°C
UN Number UN1208
Solubility Information Mixes with most organic solvents,chloroform,ether and alcohol. Insoluble in water.
Formula Weight 100.29
Percent Purity 99%
Chemical Name or Material n-Hexane-d{14}, Isotopic
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336-H361f-H372
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.