Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane-d14, for NMR, 99+ atom % D

Catalog No. AC244010010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC244010010 1 g Glass Bottle
AC244010050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC244010010 Supplier Thermo Scientific Chemicals Supplier No. 244010010
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21666-38-6 | C6H14 | 100.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21666-38-6
Molecular Formula C6H14
Molecular Weight (g/mol) 100.26
MDL Number MFCD00037562
InChI Key VLKZOEOYAKHREP-ZLKPZJALSA-N
Synonym hexane-d14, tetradecadeuterohexane, n-hexane-d 14, 2h14 hexane, 2 h?? hexane, hexane-d14, 99 atom % d
PubChem CID 140854
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecadeuteriohexane
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 g
Packaging Glass Bottle
Density 0.9600g/mL
Deuterium (D) 99.50% min.
Linear Formula D3C(CD2)4CD3
Formula Weight 100.28
Grade NMR
Chemical Name or Material n-Hexane-d14, 99.5+ atom % D
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation. Toxic to aquatic life with long lasting effects. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.