Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, MP Biomedicals™

Catalog No. ICN19522090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.5 mL
100 mL
250 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19522090 0.5 mL
ICN19522080 100 mL
MP219522083 250 mL
ICN19522091 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19522090 Supplier MP Biomedicals, Inc Supplier No. 0219522090
Add to Cart
Add to Cart

n-Hexane, ≥99%, Liquid

  • Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP. Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a reference fuel with the cetane number 100 and is employed in determining the cetane number of diesel fuels. The mechanism of the oxidation of hexadecane in the gas-phase has been described. The density, speed of sound, adiabatic compressibility and compressed liquid density of hexadecane have been determined. The performance of platinum/zeolite catalysts for the hydroisomerization of n-hexadecane has been assessed. The thermal degradation of n-hexadecane forms n-alkanes and 1-alkenes as key products.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C (Literature)
Color 1 (APHA)
Density 0.66 at 20°C (water = 1); 0.659g/mL at 25°C(Lit.)
Quantity 0.5 mL
Physical Form Liquid
Refractive Index n20/D 1.375(Lit.)
Residue after Evaporation <0 ppm
Specific Gravity 0.674
Assay Percent Range 99.9%
Vapor Pressure 124 mm Hg at 20°C 150 mm Hg at 24.8°C; 256 mmHg ( 37.7°C), 5.2 psi ( 37.7°C), ∽132 mmHg ( 20°C)(Lit.)
UN Number UN1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point 69°C (Literature)
Solubility Information Soluble in water.
Formula Weight 86.18
Chemical Name or Material n-Hexane, For ACS analysis
Show More Show Less
RTECSNumber MN9275000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.