Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hexanoyl-D-glucosamine 98.0+%, TCI America™
SDP

Catalog No. H01181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H01181G Supplier TCI America Supplier No. H01181G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19817-88-0
Molecular Formula C12H23NO6
Molecular Weight (g/mol) 277.317
MDL Number MFCD00059806
InChI Key QLZZYPWIRVQENX-LUTQBAROSA-N
Synonym 2-Deoxy-2-hexanamido-D-glucopyranose
PubChem CID 22896151
IUPAC Name N-[(2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]hexanamide
SMILES CCCCCC(=O)NC(C=O)C(C(C(CO)O)O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 277.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Hexanoyl-D-glucosamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.