Learn More
Abcam N-Hexanoyl-L-homoserine Lactone, 199.3 g/mol, >98% Purity, 10 mg
Description
- Purity: >98%
- Molecular Weight: 199.3 g/mol
- Chemical Class: Member of the N-acyl-homoserine lactone (AHL) family
- Function: Autoinducer involved in bacterial quorum sensing and regulation of gene expression in gram-negative bacteria
- Composition: 100% N-Hexanoyl-L-homoserine lactone
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N-Acyl-homoserine Lactone |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% N-Hexanoyl-L-homoserine lactone |
| Content And Storage | Store at 4°C. |
| Formulation | 100% N-Hexanoyl-L-homoserine lactone |
| Quantity | 10 mg |
| For Use With (Application) | Bacterial quorum sensing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.