Learn More
Abcam N-Hexanoyl-L-homoserine Lactone, 98% Purity, 50 mg
Description
- Molecular weight: 199.3 g/mol
- Purity: >98%
- CAS Number: 147852-83-3
- Function: Autoinducer involved in quorum sensing, a member of the N-acyl-homoserine lactone (AHL) family that regulates gene expression in gram-negative bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% N-Hexanoyl-L-homoserine lactone |
| Content And Storage | Store at 4°C. |
| Shipping Condition | Ambient |
| Target Molecule | N-Hexanoyl-L-homoserine lactone |
| Formulation | 100% N-Hexanoyl-L-homoserine lactone |
| Quantity | 50 mg |
| For Use With (Application) | Bacterial quorum sensing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.