Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxy-1,8-naphthalimide 97.0+%, TCI America™
SDP

Catalog No. H104025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H104025G 25 g
H10405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H104025G Supplier TCI America Supplier No. H104025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7797-81-1
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.192
MDL Number MFCD00065062
InChI Key KTWCUGUUDHJVIH-UHFFFAOYSA-N
Synonym n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2
PubChem CID 82263
IUPAC Name 2-hydroxybenzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 213.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxy-1,8-naphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.