Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Hydroxymethyl)nicotinamide 97.0+%, TCI America™
SDP

Catalog No. H100125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H100125G 25 g
H1001500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H100125G Supplier TCI America Supplier No. H100125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3569-99-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00010437
InChI Key JRFKIOFLCXKVOT-UHFFFAOYSA-N
PubChem CID 77116
IUPAC Name N-(hydroxymethyl)pyridine-3-carboxamide
SMILES C1=CC(=CN=C1)C(=O)NCO

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Hydroxymethyl)nicotinamide
RTECSNumber QS4476500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.