Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxyphthalimide 99.0+%, TCI America™
SDP

Catalog No. H0395100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0395100G 100 g
H039525G 25 g
H0395500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0395100G Supplier TCI America Supplier No. H0395100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 524-38-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00005891
InChI Key CFMZSMGAMPBRBE-UHFFFAOYSA-N
Synonym n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
PubChem CID 10665
IUPAC Name 2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
SMILES ON1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 100 g
Formula Weight 163.13
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxyphthalimide
EINECSNumber (5)-6724
RTECSNumber TI5200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.