Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxyurethane 85.0+%, TCI America™
SDP

Catalog No. H06181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06181G 1 g
H06185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06181G Supplier TCI America Supplier No. H06181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 589-41-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00002108
InChI Key VGEWEGHHYWGXGG-UHFFFAOYSA-N
Synonym n-hydroxyurethane, ethyl hydroxycarbamate, n-hydroxyurethan, hydroxyurethane, hydroxycarbamic acid ethyl ester, n-carbethoxyhydroxylamine, n-hydroxy ethyl carbamate, hydroxyurethan, carbamic acid, hydroxy-, ethyl ester, ethyl-n-hydroxycarbamate
PubChem CID 11510
IUPAC Name ethyl N-hydroxycarbamate
SMILES CCOC(=O)NO

Specifications

Color Yellow
Boiling Point 113°C
Quantity 1 g
Formula Weight 105.09
Percent Purity ≥85.0% (GC,N)
Physical Form Liquid
Chemical Name or Material N-Hydroxyurethane
RTECSNumber FB1750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.