Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine, 98.7%, MP Biomedicals™

Catalog No. ICN19011050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19011050 50 g
ICN19011080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19011050 Supplier MP Biomedicals, Inc Supplier No. 0219011050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-78-9
Molecular Formula C15H29NO3
Molecular Weight (g/mol) 271.40
MDL Number MFCD00021749
InChI Key BACYUWVYYTXETD-UHFFFAOYSA-N
Synonym Hamposyl L
PubChem CID 7348
IUPAC Name 2-(N-methyldodecanamido)acetic acid
SMILES CCCCCCCCCCCC(=O)N(C)CC(O)=O

Specifications

Color White
Physical Form White, waxy solid
Quantity 50 g
Assay Percent Range 98.7%
Formula Weight 271.4
Chemical Name or Material N-Lauroylsarcosine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.