Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine Sodium Salt, 94%, Spectrum™ Chemical
SDP

Catalog No. 18612033 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-033 2.5 kg Poly Bottle
18-612-030 25 g Poly Bottle
18-612-031 100 g Poly Bottle
18-612-032 500 g Poly Bottle
18-612-034 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-612-033 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L12282.5KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137-16-6
Molecular Weight (g/mol) 293.39

Specifications

CAS Max % 1
pH 7.5 to 8.5
Quantity 2.5 kg
Packaging Poly Bottle
Percent Purity 94%
Grade Ungraded
Chemical Name or Material N to Lauroylsarcosine Sodium Salt

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.