Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-2-(4-methoxyphenyl)ethylamine 95.0+%, TCI America™
SDP

Catalog No. M255120MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M255120MG 20 mg
M2551200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M255120MG Supplier TCI America Supplier No. M255120MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4091-50-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00870496
InChI Key JCMWSVNNSPUNER-UHFFFAOYSA-N
Synonym 4-Methoxy-N-methylphenethylamine, 4-[2-(Methylamino)ethyl]anisole
PubChem CID 104735
ChEBI CHEBI:75143
IUPAC Name [2-(4-methoxyphenyl)ethyl](methyl)amine
SMILES CNCCC1=CC=C(OC)C=C1

Specifications

Color Yellow
Boiling Point 83°C
Quantity 20 mg
Formula Weight 165.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-2-(4-methoxyphenyl)ethylamine
RTECSNumber SH8110000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.