Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-2-phenoxyethylamine, 98%

Catalog No. AAH5025803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5025803 1 g
AAH50258MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5025803 Supplier Thermo Scientific Chemicals Supplier No. H5025803
Add to Cart

CAS: 37421-04-8 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37421-04-8
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00085690
InChI Key GUMBNOITRNRLCS-UHFFFAOYSA-N
Synonym methyl 2-phenoxyethyl amine, n-methyl-2-phenoxyethylamine, methyl-2-phenoxy-ethyl-amine, n-methyl-2-phenoxy-ethylamin, ethylamine, n-methyl-2-phenoxy, ethanamine, n-methyl-2-phenoxy, n-methyl-n-2-phenoxyethyl amine, alpha-phenoxy-beta-methylaminoethane, acmc-1ae2y, n-methyl-2-phenoxyethylamin
PubChem CID 37732
IUPAC Name N-methyl-2-phenoxyethanamine
SMILES CNCCOC1=CC=CC=C1

Specifications

Linear Formula C6H5OCH2CH2NHCH3
Quantity 1 g
UN Number UN2735
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material N-Methyl-2-phenoxyethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
RTECSNumber KR9239990
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.