Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-3-(trifluoromethyl)benzylamine, 95%

Catalog No. AAH6388803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6388803 1 g
AAH6388806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6388803 Supplier Thermo Scientific Chemicals Supplier No. H6388803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 90390-07-1 | C9H10F3N | 189.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90390-07-1
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD04115410
InChI Key JFLPPELZYKHKQZ-UHFFFAOYSA-N
Synonym n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
PubChem CID 485414
IUPAC Name N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
SMILES CNCC1=CC(=CC=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
UN Number UN2735
Formula Weight 189.18
Percent Purity 95%
Physical Form Oil
Chemical Name or Material N-Methyl-3-(trifluoromethyl)benzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.