Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-4-[(6-methylpyrazin-2-yl)oxy]benzylamine, 95%, Thermo Scientific™

Catalog No. CC62746DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62746DA 1 g Amber Glass Bottle
CC62746CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62746DA Supplier Thermo Scientific Chemicals Supplier No. CC62746DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 912569-67-6
Molecular Formula C13H15N3O
Molecular Weight (g/mol) 229.28
MDL Number MFCD09817536
InChI Key DAHTXAIQFDPRFK-UHFFFAOYSA-N
Synonym n-methyl-4-6-methylpyrazin-2-yl oxy benzylamine, methyl 4-6-methylpyrazin-2-yl oxy phenyl methyl amine, n-methyl-1-4-6-methylpyrazin-2-yl oxy phenyl methanamine, 2-methyl-6-4-methylamino methyl phenoxy pyrazine, methyl 4-6-methylpyrazin-2-yloxy phenyl methyl amine
PubChem CID 24229697
IUPAC Name N-methyl-1-[4-(6-methylpyrazin-2-yl)oxyphenyl]methanamine
SMILES CNCC1=CC=C(OC2=NC(C)=CN=C2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 229.28
Percent Purity 95%
Chemical Name or Material N-methyl-4-[(6-methylpyrazin-2-yl)oxy]benzylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.