Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Methyl-4-phenylpyridinium Iodide, Dopaminergic Neurotoxin, Solid, 50 mg

Catalog No. 502583272
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3272 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3272 Supplier Abcam Supplier No. AB14478350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Active MPTP metabolite and dopaminergic selective neurotoxin that induces mitochondrial dysfunction.

  • Molecular Weight: 297.13 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Dopaminergic selective neurotoxin; active MPTP metabolite
  • Mechanism of Action: Interferes with oxidative phosphorylation and induces mitochondrial dysfunction; depletes cellular ATP
  • Biological Effect: Shows Parkinson effects in vivo
  • Storage: Store at -20°C

Specifications

Content And Storage Store at -20°C.
Shipping Condition Ambient
Buffer 98% MPP+ iodide
Target Molecule N-Methyl-4-phenylpyridinium Iodide
Form Solid
Formulation 98% MPP+ iodide
Quantity 50 mg
For Use With (Application) Neuroscience-Dopaminergic system study
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.