Learn More
Abcam N-Methyl-4-phenylpyridinium Iodide, Dopaminergic Selective Neurotoxin, Solid, 1 g
Description
- Dopaminergic selective neurotoxin
- Active MPTP metabolite
- Interferes with oxidative phosphorylation and induces mitochondrial dysfunction
- Depletes cellular ATP
- Shows Parkinson effects in vivo
- Purity: >98%
- Molecular weight: 297.13 Da
- Form: Solid
- CAS Number: 36913-39-0
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | N-Methyl-4-phenylpyridinium Iodide |
| Buffer | 98% MPP+ iodide |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 98% MPP+ iodide |
| Quantity | 1 g |
| For Use With (Application) | Neuroscience-Parkinson's research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.