Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-methyl-(5-pyrid-3-ylthien-2-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC66846CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC66846CB 250 mg Amber Glass Bottle
CC66846DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC66846CB Supplier Thermo Scientific Chemicals Supplier No. CC66846CB
Add to Cart

Chemical Identifiers

CAS 837376-49-5
Molecular Formula C11H12N2S
Molecular Weight (g/mol) 204.291
MDL Number MFCD11109328
InChI Key ZSTXLHXHAPANSL-UHFFFAOYSA-N
Synonym methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, 2-methylamino methyl-5-pyridin-3-ylthiophene, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1b, methyl 5-3-pyridyl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl
PubChem CID 11321674
IUPAC Name N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine
SMILES CNCC1=CC=C(S1)C2=CN=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.3
Percent Purity 97%
Chemical Name or Material N-methyl-(5-pyrid-3-ylthien-2-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.