Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-methyl-L-glutamic acid, MP Biomedicals™

Catalog No. ICN15555583 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15555583 250 mg
ICN15555591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15555583 Supplier MP Biomedicals, Inc Supplier No. 0215555583
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6753-62-4
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00065920
InChI Key LAYOOVKHWQMRIQ-WCCKRBBISA-N
Synonym n-me-glu-oh, h-l-meglu-oh hcl, n-alpha-methyl-l-glutamic acid hydrochloride, n-methyl-l-glutamic acid hydrochloride, s-2-methylamino pentanedioic acid hydrochloride
PubChem CID 56777325
IUPAC Name (2S)-2-(methylamino)pentanedioic acid;hydrochloride
SMILES CNC(CCC(=O)O)C(=O)O.Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 161.16
Physical Form Solid
Chemical Name or Material N-methyl-L-glutamic acid
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.