Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Methyl-L-isoleucine hydrochloride, 95%

Click to view available options
Quantity:
1g
5g
25g
66866-68-0
C7H16ClNO2
181.66
MFCD06801364
MMBGZHHBWVCZFK-GEMLJDPKSA-N
2s,3s-3-methyl-2-methylamino pentanoic acid hydrochloride,n-methyl-l-isoleucine hydrochloride,n-me-ile-oh,methyl l-+-isoleucine hydrochloride,n-alpha-methyl-l-isoleucine hydrochloride,n-methyl-l-isoleucine-hydrogen chloride 1/1
56777139
(2S,3S)-3-methyl-2-(methylamino)pentanoic acid;hydrochloride
CCC(C)C(C(=O)O)NC.Cl
66866-68-0
C7H16ClNO2
181.66
MFCD06801364
MMBGZHHBWVCZFK-GEMLJDPKSA-N
2s,3s-3-methyl-2-methylamino pentanoic acid hydrochloride,n-methyl-l-isoleucine hydrochloride,n-me-ile-oh,methyl l-+-isoleucine hydrochloride,n-alpha-methyl-l-isoleucine hydrochloride,n-methyl-l-isoleucine-hydrogen chloride 1/1
56777139
(2S,3S)-3-methyl-2-(methylamino)pentanoic acid;hydrochloride
CCC(C)C(C(=O)O)NC.Cl
Beilstein 3689642
Formula Weight 181.66
Physical Form Solid
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Methyl-L-isoleucine hydrochloride