Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™

Catalog No. CC49946DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC49946DA 1 g
CC49946CB 250 mg
CC49946DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC49946DA Supplier Thermo Scientific Chemicals Supplier No. CC49946DA
Add to Cart

Chemical Identifiers

CAS 361394-74-3
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
InChI Key IMIUMEQDSKHKST-UHFFFAOYSA-N
Synonym n-methyl-2-phenoxybenzylamine, n-methyl-1-2-phenoxyphenyl methanamine, n-methyl-n-2-phenoxybenzyl amine, methyl 2-phenoxyphenyl methyl amine, benzenemethanamine, n-methyl-2-phenoxy, n-methyl-2-phenoxybenzenemethanamine, n-methyl-n-2-phenoxy benzylamine, n-methyl-1-2-phenoxyphenyl ;methanamine, n-methyl-1-2-phenoxyphenyl methanamine hydrochloride
PubChem CID 23512750
IUPAC Name N-methyl-1-(2-phenoxyphenyl)methanamine
SMILES CNCC1=CC=CC=C1OC2=CC=CC=C2

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 213.28
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(2-phenoxybenzyl)amine
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.