Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(trifluoromethylthio)aniline 98.0+%, TCI America™
SDP

Catalog No. M25951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25951G 1 g
M2595200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25951G Supplier TCI America Supplier No. M25951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66476-44-6
Molecular Formula C8H8F3NS
Molecular Weight (g/mol) 207.214
InChI Key TYFGNILTFVMYGX-UHFFFAOYSA-N
PubChem CID 25013449
IUPAC Name N-methyl-N-(trifluoromethylsulfanyl)aniline
SMILES CN(C1=CC=CC=C1)SC(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 207.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-(trifluoromethylthio)aniline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.