Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-methyl-N-(trimethylsilyl)-trifluoroacetamide, MP Biomedicals™

Catalog No. ICN15567525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15567525 25 g
ICN15567505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15567525 Supplier MP Biomedicals, Inc Supplier No. 0215567525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Colorless
Density 1.075 g/mL
Quantity 25 g
Formula Weight 199.25
Physical Form Liquid
Chemical Name or Material N-methyl-N-(trimethylsilyl)-trifluoroacetamide
EINECSNumber 246-331-6
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.