CAS RN 24589-78-4
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
CAS: 24589-78-4 Molecular Formula: C6H12F3NOSi Molecular Weight (g/mol): 199.25 MDL Number: MFCD00000411 InChI Key: MSPCIZMDDUQPGJ-UHFFFAOYSA-N Synonym: mstfa,n-methyl-n-trimethylsilyl trifluoroacetamide,2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide,n-methyl-n-trimethylsilyltrifluoroacetamide,acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl,n-methyl-n-trimethylsilyl-trifluoroacetamide,acmc-209tfm PubChem CID: 32510 ChEBI: CHEBI:85064 IUPAC Name: 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide SMILES: CN(C(=O)C(F)(F)F)[Si](C)(C)C
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
CAS: 24589-78-4 Molecular Formula: C6H12F3NOSi Molecular Weight (g/mol): 199.25 MDL Number: MFCD00000411 InChI Key: MSPCIZMDDUQPGJ-UHFFFAOYSA-N Synonym: mstfa,n-methyl-n-trimethylsilyl trifluoroacetamide,2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide,n-methyl-n-trimethylsilyltrifluoroacetamide,acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl,n-methyl-n-trimethylsilyl-trifluoroacetamide,acmc-209tfm PubChem CID: 32510 ChEBI: CHEBI:85064 IUPAC Name: 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide SMILES: CN(C(=O)C(F)(F)F)[Si](C)(C)C
N-Methyl-N-trimethylsilyltrifluoroacetamide activated II, MilliporeSigma™ Supelco™
CAS: 24589-78-4 Synonym: MSTFA activated II
N-Methyl-N-(trimethylsilyl)trifluoroacetamide synthesis grade, ≥98%, MilliporeSigma™ Supelco™
MDL Number: MFCD00000411 Synonym: N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA
N-methyl-N-(trimethylsilyl)-trifluoroacetamide, MP Biomedicals™
CAS: 24589-78-4 Molecular Formula: C6H12F3NOSi Molecular Weight (g/mol): 199.25 MDL Number: MFCD00000411 InChI Key: MSPCIZMDDUQPGJ-UHFFFAOYSA-N Synonym: mstfa,n-methyl-n-trimethylsilyl trifluoroacetamide,2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide,n-methyl-n-trimethylsilyltrifluoroacetamide,acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl,n-methyl-n-trimethylsilyl-trifluoroacetamide,acmc-209tfm PubChem CID: 32510 ChEBI: CHEBI:85064 IUPAC Name: 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide SMILES: CN(C(=O)C(F)(F)F)[Si](C)(C)C
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent for Silylations, MilliporeSigma™ Supelco™
MDL Number: MFCD00000411 Synonym: N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1 trimethylchlorosilane, ≥97%, MilliporeSigma™ Supelco™
Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).
LiChropur™ N-Methyl-N-trimethylsilyltrifluoroacetamide Activated I, For GC Derivatization, MilliporeSigma™ Supelco™
Activated with Ethanethiol and Ammonium iodide
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 98.5%, MilliporeSigma™ Supelco™
N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a silylating reagent.
N-Methyl-N-trimethylsilyltrifluoroacetamide 95.0+%, TCI America™
CAS: 24589-78-4 Molecular Formula: C6H12F3NOSi Molecular Weight (g/mol): 199.25 MDL Number: MFCD00000411 InChI Key: MSPCIZMDDUQPGJ-UHFFFAOYSA-N Synonym: mstfa,n-methyl-n-trimethylsilyl trifluoroacetamide,2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide,n-methyl-n-trimethylsilyltrifluoroacetamide,acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl,n-methyl-n-trimethylsilyl-trifluoroacetamide,acmc-209tfm PubChem CID: 32510 ChEBI: CHEBI:85064 IUPAC Name: 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide SMILES: CN(C(=O)C(F)(F)F)[Si](C)(C)C