Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(trimethylsilyl)trifluoroacetamide synthesis grade, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111018738 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8738 10 x 1 mL
11-101-9191 5 mL
11-101-8265 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8738 Supplier MilliporeSigma Supelco Supplier No. 39486610X1ML
Add to Cart
Add to Cart

N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a derivatizing reagent. It can be used as alternatives to diazomethane.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00000411
Synonym N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA

Specifications

Density 1.075 g/mL (at 25°C)
CAS 24589-78-4
Boiling Point 130°C to 132°C
Flash Point 26°C - Closed Cup
Packaging Ampule
Linear Formula CF3 CON(CH3)Si(CH3)3
Quantity 10 x 1 mL
Vapor Density >1 (vs. air)
Formula Weight 199.25
Vapor Pressure 8.8 mmHg (27°C)
Percent Purity ≥98%
Grade Synthesis Grade
Show More Show Less
RTECSNumber AC9800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.