Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmaleic hydrazide, 97%

Catalog No. aab2097514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2097514 25 g
AAB2097506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2097514 Supplier Thermo Scientific Chemicals Supplier No. B2097514
Only null left
Add to Cart

CAS: 5436-01-1 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5436-01-1
MDL Number MFCD00234108

Specifications

Melting Point 216°C to 219°C
Molecular Formula C5H6N2O2
Quantity 25 g
Synonym 3-hydroxy-1-methylpyridazin-6 1h-one, 1-methyl-3,6-pyridazinedione, n-methyl-3,6-pyridazinedione, n-methylmaleic hydrazide, 6-hydroxy-2-methylpyridazin-3 2h-one, n-methyl maleic hydrazide, 1-methyl-3,6-1h,2h pyridazinedione, 1-methyl-1,2-dihydropyridazine-3,6-dione, 6-hydroxy-2-methylpyridazin-3-one, 1-methyl-1,2-dihydro-3,6-pyridazinedione
InChI Key UAECOHJYXUJDOF-UHFFFAOYSA-N
SMILES CN1C(=O)C=CC(=O)N1
IUPAC Name 2-methyl-1H-pyridazine-3,6-dione
Molecular Weight (g/mol) 126.12
PubChem CID 95157
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material N-Methylmaleic hydrazide
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.