Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ n-Myc Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111454 Supplier Invitrogen™ Supplier No. PA5111454
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEKRRSSSNT KAVTTFTITV RPKNAALGPG RAQSSELILK RCLPIHQQHN YAA Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 87%.

N-Myc protein is member of the MYC family of transcription factor proteins with a basic helix-loop-helix (bHLH) domain. N-Myc is similarly regulated and functionally complementary to C-Myc in the context of various cellular growth and differentiation processes. MYCN gene is considered the hallmark of neuroblastoma, amplification and overexpression of the MYCN proto-oncogene occurs in approximately 20% of neuroblastomas and is associated with advanced stage disease, rapid tumor progression, and poor prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen n-Myc
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYCN
Gene Accession No. P04198
Gene Alias Avian myelocytomatosis viral (v-myc) related oncogene neuroblastoma derived (Nmyc); Avian myelocytomatosis viral (v-myc) related oncogene, neuroblastoma derived (Nmyc); BHLHE37; Class E basic helix-loop-helix protein 37; c-nmyc; fb57a02; MODED; Mycn; mycn protein; MYCN proto-oncogene, bHLH transcription factor; neuroblastoma MYC oncogene; neuroblastoma myc-related oncogene 1; neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene; Nmuc1; NMYC; N-myc; N-myc protein; N-myc proto-oncogene protein; Nmyc1; Nmyc-1; N-myc-like protein; ODED; oncogene NMYC; oncogene pp65/67; pp65/67; v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian); wu:fb57a02; zgc:85706
Gene Symbols MYCN
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human n-Myc. Recombinant protein control fragment (Product #RP-106776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.