Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid, MP Biomedicals™

Catalog No. ICN10092783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10092783 250 g
ICN10092780 100 g
ICN10092791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10092783 Supplier MP Biomedicals, Inc Supplier No. 0210092783
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-Bis(2-hydroxyethyl)taurine, N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 0.9961
Formula Weight 239.23
Chemical Name or Material N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.