Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(3-aminopropyl)ethylenediamine 98.0+%, TCI America™
SDP

Catalog No. B195225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B195225ML 25 mL
B1952500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B195225ML Supplier TCI America Supplier No. B195225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10563-26-5
Molecular Formula C8H22N4
Molecular Weight (g/mol) 174.292
MDL Number MFCD00008210
InChI Key RXFCIXRFAJRBSG-UHFFFAOYSA-N
Synonym n,n'-bis 3-aminopropyl ethylenediamine, 1,5,8,12-tetraazadodecane, n1,n1'-ethane-1,2-diyl bis propane-1,3-diamine, 1,3-propanediamine, n,n-1,2-ethanediylbis, n,n'-bis 3-aminopropyl-1,2-diaminoethane, n,n'-di 3-aminopropyl-1,2-ethylenediamine, n,n'-bis gamma-aminopropyl diaminoethane, 1,3-propanediamine, n,n-ethylenebis, 1,2-bis 3-aminopropylamino ethane, unii-0exw8894xx
PubChem CID 25378
ChEBI CHEBI:39476
IUPAC Name N'-[2-(3-aminopropylamino)ethyl]propane-1,3-diamine
SMILES C(CN)CNCCNCCCN

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
UN Number 2927
Formula Weight 174.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Bis(3-aminopropyl)ethylenediamine
EINECSNumber (7)-0005
RTECSNumber TX8032950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.