Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(4-biphenylyl)aniline 98.0+%, TCI America™
SDP

Catalog No. B50861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50861G 1 g
B50865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50861G Supplier TCI America Supplier No. B50861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122215-84-3
Molecular Formula C30H23N
Molecular Weight (g/mol) 397.521
MDL Number MFCD20483034
InChI Key VXKVZTGWNITVPY-UHFFFAOYSA-N
PubChem CID 15100105
IUPAC Name N,4-diphenyl-N-(4-phenylphenyl)aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N(C3=CC=CC=C3)C4=CC=C(C=C4)C5=CC=CC=C5

Specifications

Melting Point 171°C
Color White
Quantity 1 g
Synonym N-([1,1′C-Biphenyl]-4-yl)-N-phenyl-[1,1′C-biphenyl]-4-amine
Formula Weight 397.52
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-biphenylyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.