Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(salicylidene)ethylenediamine 99.0+%, TCI America™
SDP

Catalog No. H019925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H019925G 25 g
H0199500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H019925G Supplier TCI America Supplier No. H019925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-93-9
Molecular Formula C16H16N2O2
Molecular Weight (g/mol) 268.32
MDL Number MFCD00002244
InChI Key RQHVNNWVDLRULK-XSYHWHKQSA-N
Synonym salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine
PubChem CID 5464119
IUPAC Name (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O

Specifications

Melting Point 127°C
Color Yellow
Quantity 25 g
Formula Weight 268.32
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(salicylidene)ethylenediamine
EINECSNumber (3)-2919
RTECSNumber SL3780000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.