Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di-o-tolylthiourea, 98%, Thermo Scientific™

Catalog No. AAL0363014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0363014 25 g
AAL0363006 5 g
AAL0363022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0363014 Supplier Thermo Scientific Chemicals Supplier No. L0363014

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-97-3
Molecular Formula C15H16N2S
Molecular Weight (g/mol) 256.37
MDL Number MFCD00025922
InChI Key KWPNNZKRAQDVPZ-UHFFFAOYSA-N
Synonym di-o-tolylthiourea, 1,3-di o-tolyl thiourea, 1,3-di-o-tolylthiourea, n,n'-di-o-tolylthiourea, 1,3-di-o-tolyl-2-thiourea, di-o-toluylthiourea, 1,3-bis o-tolyl thiourea, n,n'-bis 2-methylphenyl thiourea, 2,2'-dimethylthiocarbanilide, usaf ek-1651
PubChem CID 688028
IUPAC Name 1,3-bis(2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(=S)NC1=CC=CC=C1C

Specifications

Melting Point 153°C to 155°C
Quantity 25 g
Beilstein 2125770
Formula Weight 256.37
Percent Purity 98%
Chemical Name or Material N,N'-Di-o-tolylthiourea
EINECSNumber 205-309-6
RTECSNumber FE0700000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.