Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di-tert-butylcarbodiimide 98.0+%, TCI America™
SDP

Catalog No. D534725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D534725ML 25 mL
D53475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D534725ML Supplier TCI America Supplier No. D534725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 691-24-7
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.257
MDL Number MFCD00010231
InChI Key IDVWLLCLTVBSCS-UHFFFAOYSA-N
PubChem CID 69650
IUPAC Name N,N'-ditert-butylmethanediimine
SMILES CC(C)(C)N=C=NC(C)(C)C

Specifications

Color Colorless
Boiling Point 50°C
Quantity 25 mL
UN Number 1993
Formula Weight 154.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Di-tert-butylcarbodiimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.