Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dibenzoylhydrazine 95.0+%, TCI America™
SDP

Catalog No. D015025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D015025G 25 g
D01505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D015025G Supplier TCI America Supplier No. D015025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 787-84-8
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.262
MDL Number MFCD00003068
InChI Key GRRIYLZJLGTQJX-UHFFFAOYSA-N
Synonym dibenzoylhydrazine, n,n'-dibenzoylhydrazine, 1,2-dibenzoylhydrazine, benzoic acid, 2-benzoylhydrazide, n2-benzoylbenzoic acid hydrazide, hydrazine, 1,2-dibenzoyl, n,n-dibenzoylhydrazine, nn'-dibenzoylhydrazine, c6h5-co-nh-nh-co-c6h5, benzoic acid,2-benzoylhydrazide
PubChem CID 13085
ChEBI CHEBI:38453
IUPAC Name N'-benzoylbenzohydrazide
SMILES C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=CC=C2

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 240.26
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dibenzoylhydrazine
RTECSNumber MV2085000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.