Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibenzylglycine Ethyl Ester 98.0+%, TCI America™
SDP

Catalog No. D316525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D316525G 25 g
D31655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D316525G Supplier TCI America Supplier No. D316525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77385-90-1
Molecular Formula C18H21NO2
Molecular Weight (g/mol) 283.371
InChI Key AFMDCFOWHWNQBP-UHFFFAOYSA-N
Synonym Di-Bzl-Gly-OEt
PubChem CID 3553452
IUPAC Name ethyl 2-(dibenzylamino)acetate
SMILES CCOC(=O)CN(CC1=CC=CC=C1)CC2=CC=CC=C2

Specifications

Melting Point 56°C
Color White
Quantity 25 g
Formula Weight 283.37
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylglycine Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.