Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibenzylhydroxylamine 98.0+%, TCI America™
SDP

Catalog No. D2004500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2004500G 500 g
D200425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2004500G Supplier TCI America Supplier No. D2004500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 621-07-8
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00004772
InChI Key GXELTROTKVKZBQ-UHFFFAOYSA-N
Synonym dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl
PubChem CID 69297
IUPAC Name N,N-dibenzylhydroxylamine
SMILES ON(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 124°C
Color White
Quantity 500 g
Formula Weight 213.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylhydroxylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.