Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibutylaniline 98.0+%, TCI America™
SDP

Catalog No. D0222100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0222100ML 100 mL
D022225ML 25 mL
D0222500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0222100ML Supplier TCI America Supplier No. D0222100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-29-6
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00035799
InChI Key FZPXKEPZZOEPGX-UHFFFAOYSA-N
Synonym benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl
PubChem CID 61154
IUPAC Name N,N-dibutylaniline
SMILES CCCCN(CCCC)C1=CC=CC=C1

Specifications

Boiling Point 266°C
Quantity 100 mL
Formula Weight 205.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dibutylaniline
EINECSNumber (3)-0129
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.