Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dibutylquinacridone 98.0+%, TCI America™
SDP

Catalog No. D47801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47801G 1 g
D4780200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47801G Supplier TCI America Supplier No. D47801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99762-80-8
Molecular Formula C28H28N2O2
Molecular Weight (g/mol) 424.54
MDL Number MFCD12407159
InChI Key MYFHGRHNRNCMHV-UHFFFAOYSA-N
Synonym 5,12-Dibutyl-5,12-dihydroquino[2,3-b]acridine-7,14-dione
PubChem CID 13432652
IUPAC Name 5,12-dibutyl-5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
SMILES CCCCN1C2=CC=CC=C2C(=O)C2=CC3=C(C=C12)C(=O)C1=CC=CC=C1N3CCCC

Specifications

Melting Point 265°C
Quantity 1 g
Formula Weight 424.54
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dibutylquinacridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.