Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dicyclohexyl-4-morpholinecarboxamidine 98.0+%, TCI America™
SDP

Catalog No. D379225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D379225G 25 g
D37925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D379225G Supplier TCI America Supplier No. D379225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4975-73-9
Molecular Formula C17H31N3O
Molecular Weight (g/mol) 293.455
MDL Number MFCD00063252
InChI Key OZNYZQOTXQSUJM-UHFFFAOYSA-N
Synonym n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide
PubChem CID 21079
IUPAC Name N,N'-dicyclohexylmorpholine-4-carboximidamide
SMILES C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 25 g
Formula Weight 293.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dicyclohexyl-4-morpholinecarboxamidine
RTECSNumber LQ4566500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.