Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-1,4-phenylenediamine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D051925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051925G 25 g
D05195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D051925G Supplier TCI America Supplier No. D051925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16713-15-8
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
MDL Number MFCD00060227
InChI Key NHPCFKHZILUMDE-UHFFFAOYSA-N
Synonym 4-Amino-N,N-diethylaniline Dihydrochloride
PubChem CID 19799983
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;dihydrochloride
SMILES CCN(CC)C1=CC=C(C=C1)N.Cl.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 237.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-1,4-phenylenediamine Dihydrochloride
EINECSNumber (3)-0243
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.